missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-3-Methyl-2-butanol, 99%

Product Code. p-7045106
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 11337426

Brand: Thermo Scientific Alfa Aesar L09431.MC

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1517-66-4 | C5H12O | 88.15 g/mol

It is an pharmaceutical and organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an pharmaceutical and organic intermediate.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Store in cool dry condition. Incompatible with strong oxidizing agents. Ensure ventilation. Protect from electrostatic charges.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1517-66-4
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00064271
InChI Key MXLMTQWGSQIYOW-UHFFFAOYNA-N
Synonym s-+-3-methyl-2-butanol, 2s-3-methylbutan-2-ol, 2-butanol, 3-methyl-, 2s, +-3-methyl-2-butanol, 3-methyl-2-butanol #, s-3-methyl-2-butanol, s-3-methyl-butan-2-ol, unii-2nk7o363q6 component, unii-93ff0f303r component
PubChem CID 6999784
IUPAC Name (2S)-3-methylbutan-2-ol
SMILES CC(C)C(C)O

Specifications

Density 0.81
Boiling Point 112°C
Flash Point 26°C (78°F)
Refractive Index 1.409
Quantity 100 mg
UN Number UN1105
Beilstein 1718801
Solubility Information Soluble in alcohol. Insoluble in water.
Optical Rotation +5° (Neat)
Formula Weight 88.15
Percent Purity 99%
Chemical Name or Material (S)-(+)-3-Methyl-2-butanol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.