accessibility menu, dialog, popup

UserName

CAS RN 1517-66-4

H3CCH3CH3HO

CAS RN 1517-66-4

IUPAC Name: 3-methylbutan-2-ol
Synonyms: 3-methyl-2-butanol
Molecular Weight (g/mol): 88.15
Molecular Formula: C5H12O
InChi Key: MXLMTQWGSQIYOW-UHFFFAOYNA-N
SMILES: CC(C)C(C)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results