missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-(Boc-amino)butyric acid, 95%, Thermo Scientific™

Product Code. p-7034383
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15478577 250 mg 250mg
15468577 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15478577 Supplier Thermo Scientific Alfa Aesar Supplier No. H52066.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
(S)-3-(Boc-amino)butyric acid is used for the syntheses of aza-analogues of macrosphelides, synthesis of hybrid peptides with 12/10-helix, and in the synthesis of potentially pharmaceutically active products for the treatment of auto-immune and inflammatory diseases.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 158851-30-0
Molecular Formula C9H17NO4
Molecular Weight (g/mol) 203.238
MDL Number MFCD00270345
InChI Key PYNDHEONPQYIAN-LURJTMIESA-N
Synonym boc-l-beta-homoalanine, s-3-tert-butoxycarbonyl amino butanoic acid, boc-beta-hoala-oh, boc-beta-homoala-oh, s-n-boc-3-aminobutyric acid, s-3-tert-butoxycarbonylamino butanoic acid, 3s-3-tert-butoxycarbonyl amino butanoic acid, 3s-3-2-methylpropan-2-yl oxycarbonylamino butanoic acid, butanoic acid, 3-1,1-dimethylethoxy carbonyl amino-, 3s, boc--homoalanine
PubChem CID 5706671
IUPAC Name (3S)-3-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
SMILES CC(CC(=O)O)NC(=O)OC(C)(C)C

Specifications

Quantity 250 mg
Beilstein 6800577
Solubility Information Slightly soluble in water.
Formula Weight 203.24
Percent Purity 95%
Chemical Name or Material (S)-3-(Boc-amino)butyric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.