accessibility menu, dialog, popup

UserName

CH3(S)HOONHOOCH3CH3H3C

CAS RN 158851-30-0

IUPAC Name: (3S)-3-{[(tert-butoxy)carbonyl]amino}butanoic acid
Synonyms: (3S)-3-[(tert-butoxycarbonyl)amino]butanoic acid
Molecular Weight (g/mol): 203.24
Molecular Formula: C9H17NO4
InChi Key: PYNDHEONPQYIAN-LURJTMIESA-N
SMILES: C[C@@H](CC(O)=O)NC(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results