Learn More
(S)-3-Aminopiperidin-2-one Hydrochloride, TRC
Product Code. 30006296
Change view
Click to view available options
Quantity:
50 mg
100 mg
1 g
Unit Size:
100mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006296 50 mg 50mg
30004720 1 g 1g
17974535 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30006296 Supplier Toronto Research Chemicals Supplier No. TRCA62794050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 42538-31-8
Molecular Formula C5H10N2O.ClH
Molecular Weight (g/mol) 150.607
Synonym 2-Piperidinone, 3-amino-, monohydrochloride, (3S)-, 2-Piperidinone, 3-amino-, monohydrochloride, (S)-, (3S)-3-Amino-2-oxohexahydropyridine hydrochloride, (3S)-3-Aminopiperidin-2-one hydrochloride, (S)-3-Aminopiperidin-2-one hydrochloride, (S)-3-Aminotetrahydropyridin-2-one hydrochloride, L-Ornithine-1,5-lactam Hydrochloride
IUPAC Name (3S)-3-aminopiperidin-2-one;hydrochloride
SMILES Cl.N[C@H]1CCCNC1=O
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C5H10N2O.ClH/c6-4-2-1-3-7-5(4)8;/h4H,1-3,6H2,(H,7,8);1H/t4-;/m0./s1
Formula Weight 150.056
Percent Purity >95%
Chemical Name or Material (S)-3-Aminopiperidin-2-one Hydrochloride
Grade HPLC