CAS RN 42538-31-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(S)-3-Aminopiperidin-2-one Hydrochloride, TRC
CAS: 42538-31-8 Molecular Formula: C5H10N2O.ClH Molecular Weight (g/mol): 150.607 Synonym: 2-Piperidinone, 3-amino-, monohydrochloride, (3S)-,2-Piperidinone, 3-amino-, monohydrochloride, (S)-,(3S)-3-Amino-2-oxohexahydropyridine hydrochloride,(3S)-3-Aminopiperidin-2-one hydrochloride,(S)-3-Aminopiperidin-2-one hydrochloride,(S)-3-Aminotetrahydropyridin-2-one hydrochloride,L-Ornithine-1,5-lactam Hydrochloride IUPAC Name: (3S)-3-aminopiperidin-2-one;hydrochloride SMILES: Cl.N[C@H]1CCCNC1=O