missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-Aminobutan-1-ol, TRC

Product Code. 30134505
Change view
Click to view available options
Quantity:
100 mg
2.5 g
5 g
Unit Size:
100mg
2.5g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134505 100 mg 100mg
30133435 2.5 g 2.5g
30008973 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134505 Supplier Toronto Research Chemicals Supplier No. TRCA601888100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61477-39-2
Molecular Formula C4 H11 N O
Molecular Weight (g/mol) 89.14
Synonym (3S)-3-Amino-1-butanol, (3S)-3-Aminobutan-1-ol
IUPAC Name (3S)-3-aminobutan-1-ol
SMILES C[C@H](N)CCO

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C4H11NO/c1-4(5)2-3-6/h4,6H,2-3,5H2,1H3/t4-/m0/s1
Formula Weight 89.0841
Chemical Name or Material (S)-3-Aminobutan-1-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.