CAS RN 61477-39-2
CAS RN 61477-39-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(S)-3-Aminobutan-1-ol, TRC
CAS: 61477-39-2 Molecular Formula: C4 H11 N O Molecular Weight (g/mol): 89.14 Synonym: (3S)-3-Amino-1-butanol,(3S)-3-Aminobutan-1-ol IUPAC Name: (3S)-3-aminobutan-1-ol SMILES: C[C@H](N)CCO