missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-Diphenylphosphino-1-phenylethylamine, 97+%

Product Code. 15455726
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15455726 250 mg 250mg
15496868 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15455726 Supplier Thermo Scientific Alfa Aesar Supplier No. H60404.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1103533-85-2 | C20H20NP | 305.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1103533-85-2
Molecular Formula C20H20NP
Molecular Weight (g/mol) 305.36
MDL Number MFCD17013989
InChI Key OTNYDYNCOCRMDG-GNLPSFAGNA-N
Synonym s-2-diphenylphosphino-1-phenylethanamine, s-2-diphenylphosphino-1-phenylethylamine, 2s-2-amino-2-phenylethyl diphenylphosphane, alphachiron 1112421a940, s-beta-aminophenethyl diphenylphosphine, s-2-diphenylphosphanyl-1-phenylethan-1-amine, 1s-2-diphenylphosphanyl-1-phenylethan-1-amine, s-2-diphenylphosphino-1-phenylethylamine,min
PubChem CID 70861903
IUPAC Name (1S)-2-diphenylphosphanyl-1-phenylethanamine
SMILES N[C@H](CP(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 305.35
Percent Purity ≥97%
Chemical Name or Material (S)-2-Diphenylphosphino-1-phenylethylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.