accessibility menu, dialog, popup

UserName

CAS RN 1103533-85-2

IUPAC Name: (2-amino-2-phenylethyl)diphenylphosphane
Synonyms: (2-amino-2-phenylethyl)diphenylphosphane
Molecular Weight (g/mol): 305.36
Molecular Formula: C20H20NP
InChi Key: OTNYDYNCOCRMDG-UHFFFAOYNA-N
SMILES: NC(CP(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results