Learn More
Tocris Bioscience™ Ro 64-5229
GREENER_CHOICE
Product Code. 18024602
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18024602 10 mg 10mg
18044382 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18024602 Supplier Tocris Bioscience™ Supplier No. 2913/10
Selective, non-competitive mGlu2 antagonist
Selective, non-competitive mGlu2 antagonist. Inhibits GTPγ35S binding to mGlu2-containing membranes (IC50 = 0.11 μM).
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 246852-46-0
Molecular Formula C17H19Cl2N3O
Specifications
Quantity 10 mg
Formula Weight 352.26
Purity >99%
Molecular Weight (g/mol) 352.259
Chemical Name or Material Ro 64-5229