missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Ro 08-2750
GREENER_CHOICE

Product Code. 18004852
Click to view available options
Quantity:
1 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
This item is not returnable. View return policy

Product Code. 18004852

Brand: Tocris Bioscience™ 2272/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Inhibits NGF binding to p75NTR and TrkA

Non-peptide inhibitor of NGF that binds the NGF dimer (KD ∽ 1 μM) possibly causing a conformational change. Selectively inhibits binding of NGF to p75NTR at submicromolar concentrations, and to both p75NTR and TrkA at concentrations > 5 μM. Prevents NGF-induced apoptosis in SK-N-MC cells. Suppresses cancer cell proliferation via regulation of Hippo signaling aand YAP phosphorylation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37854-59-4
Molecular Formula C13H10N4O3

Specifications

Quantity 50 mg
Formula Weight 270.24
Purity >96%
Molecular Weight (g/mol) 270.248
Chemical Name or Material Ro 08-2750
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.