CAS RN 37854-59-4
CAS RN 37854-59-4
IUPAC Name:
7,10-dimethyl-2,4-dioxo-2H,3H,4H,10H-benzo[g]pteridine-8-carbaldehyde
Synonyms:
7,10-dimethyl-2,4-dioxo-3H-benzo[g]pteridine-8-carbaldehyde
Molecular Weight (g/mol):
270.25
Molecular Formula:
C13H10N4O3
InChi Key:
JDEMVNYMYPJJIM-UHFFFAOYSA-N
SMILES:
CN1C2=CC(C=O)=C(C)C=C2N=C2C(=O)NC(=O)N=C12