missing translation for 'onlineSavingsMsg'
Learn More

Resiquimod, TRC

Product Code. 17943315
Change view
Click to view available options
Quantity:
5 mg
50 mg
250 mg
Unit Size:
250mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17943315 5 mg 5mg
17943305 50 mg 50mg
17911395 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17943315 Supplier Toronto Research Chemicals Supplier No. TRCR1446805MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144875-48-9
Molecular Formula C17 H22 N4 O2
Molecular Weight (g/mol) 314.38
Synonym 1-[4-Amino-2-(ethoxymethyl)-1H-imidazo[4,5-c]quinolin-1-yl]-2-methylpropan-2-ol, 4-Amino-2-(ethoxymethyl)-1-(2-hydroxyisobutyl)imidazo[4,5-c]quinoline, 4-Amino-2-ethoxymethyl-α,α-dimethyl-1H-imidazo[4,5-c]quinoline-1-ethanol, R 848, Resiquimod, S 28463
IUPAC Name 1-[4-amino-2-(ethoxymethyl)imidazo[4,5-c]quinolin-1-yl]-2-methylpropan-2-ol
SMILES CCOCc1nc2c(N)nc3ccccc3c2n1CC(C)(C)O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C17H22N4O2/c1-4-23-9-13-20-14-15(21(13)10-17(2,3)22)11-7-5-6-8-12(11)19-16(14)18/h5-8,22H,4,9-10H2,1-3H3,(H2,18,19)
Formula Weight 314.1743
Chemical Name or Material Resiquimod
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.