missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-170 of Human UBE2G1 The Recombinant Human UBE2G1 Protein is derived from E. coli. The Recombinant Human UBE2G1 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Concentration | 0.5mg/mL |
| For Use With (Application) | SDS-PAGE |
| Formulation | Phosphate buffered saline (pH7.4) containing, 30% glycerol, 1mM DTT |
| Gene ID (Entrez) | 7326 |
| Molecular Weight (g/mol) | TMW: 21.9kDa |
| Quantity | 0.1 mg |
| Storage Requirements | Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. |
| Gene Alias | EC 6.3.2.19, UBC7, UBE2GE217K, Ubiquitin carrier protein G1, ubiquitin-conjugating enzyme E2 G1, ubiquitin-conjugating enzyme E2G 1 (homologous to C. elegans UBC7), ubiquitin-conjugating enzyme E2G 1 (UBC7 homolog, C. elegans), ubiquitin-conjugating enzyme E2G 1 (UBC7 homolog, yeast), Ubiquitin-protein ligase G1 |
| Common Name | UBE2G1 |
| Conjugate | Unconjugated |
| Show More |
For research use only and is not approved for use in humans or in clinical diagnosis
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?