missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human KNG1 / BDK / kininogen-1 Protein
A DNA sequence encoding the Homo sapiens (Human) KNG1 / BDK / kininogen-1, was expressed in the hosts and tags indicated.
160.00 EUR - 2090.00 EUR
Specifications
Accession Number | NP_000884.1 |
---|---|
Gene ID (Entrez) | 3827 |
Name | KNG1 / BDK / kininogen-1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16006601
|
enQuireBio™
QP8809-EC-100UG |
100 μg |
463.00 EUR
100µg |
Estimated Shipment: 10-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16026601
|
enQuireBio™
QP8809-EC-1MG |
1 mg |
2090.00 EUR
1mg |
Estimated Shipment: 10-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16016601
|
enQuireBio™
QP8809-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16036601
|
enQuireBio™
QP8809-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16046601
|
enQuireBio™
QP8809-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16056601
|
enQuireBio™
QP8809-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000884.1 | |
KNG1 / BDK / kininogen-1 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
3827 | |
Research Use Only | |
KNG1 | |
Recombinant Protein | |
E. coli | |
SSRIGEIKEE TTVSPPHTSM APAQDEERDS GKEQGHTRRH DWGHEKQRKH NLGHGHKHER DQGHGHQRGH GLGHGHEQQH GLGHGHKFKL DDDLEHQGGH VLDHGHKHKH GHGHGKHKNK GKKNGKHNGW KTEHLASSSE DSTTPSAQTQ EKTEGPTPIP SLAKPGVTVT FSDFQDSDLI ATMMPPISPA PIQSDDDWIP DIQIDPNGLS FNPISDFPDT TSPKCPGRPW KSVSEINPTT QMKESYYFDL TDGLS | |
Greater than 90% as determined by SDS-PAGE. |