missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human KNG1 / BDK / kininogen-1 Protein
A DNA sequence encoding the Homo sapiens (Human) KNG1 / BDK / kininogen-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8809-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000884.1 | |
KNG1 / BDK / kininogen-1 Protein | |
Research Use Only | |
KNG1 | |
Recombinant Protein | |
E. coli | |
SSRIGEIKEE TTVSPPHTSM APAQDEERDS GKEQGHTRRH DWGHEKQRKH NLGHGHKHER DQGHGHQRGH GLGHGHEQQH GLGHGHKFKL DDDLEHQGGH VLDHGHKHKH GHGHGKHKNK GKKNGKHNGW KTEHLASSSE DSTTPSAQTQ EKTEGPTPIP SLAKPGVTVT FSDFQDSDLI ATMMPPISPA PIQSDDDWIP DIQIDPNGLS FNPISDFPDT TSPKCPGRPW KSVSEINPTT QMKESYYFDL TDGLS | |
Greater than 90% as determined by SDS-PAGE. |
3827 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |