missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human KNG1 / BDK / kininogen-1 Protein

A DNA sequence encoding the Homo sapiens (Human) KNG1 / BDK / kininogen-1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8809-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16046601

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_000884.1
KNG1 / BDK / kininogen-1 Protein
Research Use Only
KNG1
Recombinant Protein
E. coli
SSRIGEIKEE TTVSPPHTSM APAQDEERDS GKEQGHTRRH DWGHEKQRKH NLGHGHKHER DQGHGHQRGH GLGHGHEQQH GLGHGHKFKL DDDLEHQGGH VLDHGHKHKH GHGHGKHKNK GKKNGKHNGW KTEHLASSSE DSTTPSAQTQ EKTEGPTPIP SLAKPGVTVT FSDFQDSDLI ATMMPPISPA PIQSDDDWIP DIQIDPNGLS FNPISDFPDT TSPKCPGRPW KSVSEINPTT QMKESYYFDL TDGLS
Greater than 90% as determined by SDS-PAGE.
3827
500 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers