Learn More
enQuireBio™ Recombinant Human KNG1 / BDK / kininogen-1 Protein
A DNA sequence encoding the Homo sapiens (Human) KNG1 / BDK / kininogen-1, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP8809-ec-1mg
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 16026601
  • 2090.00 EUR / 1mg
Estimated Shipment: 19-07-2024
Specifications
NP_000884.1
KNG1 / BDK / kininogen-1 Protein
Research Use Only
KNG1
Recombinant Protein
E. coli
SSRIGEIKEE TTVSPPHTSM APAQDEERDS GKEQGHTRRH DWGHEKQRKH NLGHGHKHER DQGHGHQRGH GLGHGHEQQH GLGHGHKFKL DDDLEHQGGH VLDHGHKHKH GHGHGKHKNK GKKNGKHNGW KTEHLASSSE DSTTPSAQTQ EKTEGPTPIP SLAKPGVTVT FSDFQDSDLI ATMMPPISPA PIQSDDDWIP DIQIDPNGLS FNPISDFPDT TSPKCPGRPW KSVSEINPTT QMKESYYFDL TDGLS
Greater than 90% as determined by SDS-PAGE.
3827
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol