missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
A recombinant protein with a C-terminalHis-tag and corresponding to the amino acids 210-297 of Human CD20. The Recombinant Human CD20 Protein is derived from Baculovirus. The Recombinant Human CD20 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Concentration | 0.5 mg/ml |
| For Use With (Application) | SDS-PAGE |
| Formulation | Phosphate Buffered Saline (pH 7.4) containing 10% glycerol. |
| Gene ID (Entrez) | 931 |
| Molecular Weight (g/mol) | 11.1 kDa |
| Quantity | 10 μg |
| Source | Baculovirus |
| Storage Requirements | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Regulatory Status | RUO |
| Gene Symbol | MS4A1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?