Learn More
R&D Systems™ Rat TrkB (NP_036863) VersaClone cDNA
Specifications
Specifications
| Accession Number | (NP_036863 |
| Gene ID (Entrez) | 4915 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date |
| Species | Rat |
| Product Line | R&D Systems |
| Quantity | 10 μg |