missing translation for 'onlineSavingsMsg'
Learn More

RASGRP2 Antibody, Novus Biologicals™

Product Code. 18214730 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18214730 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18214730 Supplier Novus Biologicals Supplier No. NBP133315
  • ONLINE EXCLUSIVE PRICE
    629.00 EUR
    594.30 EUR / 100µL
    Save 34.70 EUR
    5% Off
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RASGRP2 Polyclonal specifically detects RASGRP2 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RASGRP2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.0mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunoprecipitation 1:100-1:500, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Gene Alias Calcium and DAG-regulated guanine nucleotide exchange factor I, CALDAG-GEFI, CDC25Lcalcium and diacylglycerol-regulated guanine nucleotide exchange factor I, Cdc25-like protein, F25B3.3 kinase-like protein, guanine exchange factor MCG7, hCDC25L, MCG7, RAS guanyl nucleotide-releasing protein 2, RAS guanyl releasing protein 2 (calcium and DAG-regulated), RAS guanyl-releasing protein 2
Gene Symbols RASGRP2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human RASGRP2. The exact sequence is proprietary.
Molecular Weight of Antigen 69 kDa
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10235
Test Specificity Tested in human. Not yet tested in any other species.
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.