Learn More
rac Salsolinol Hydrochloride, TRC
Product Code. 30428274
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30428274 1 g 1g
30410335 5 g -
30422240 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30428274 Supplier Toronto Research Chemicals Supplier No. TRCS1000001G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 79923-51-6
Molecular Formula C10 H13 N O2 . Cl H
Molecular Weight (g/mol) 215.68
IUPAC Name 1-methyl-1,2,3,4-tetrahydroisoquinoline-6,7-diol;hydrochloride
SMILES Cl.CC1NCCc2cc(O)c(O)cc12
Specifications
Quantity 1 g
InChI Formula InChI=1S/C10H13NO2.ClH/c1-6-8-5-10(13)9(12)4-7(8)2-3-11-6;/h4-6,11-13H,2-3H2,1H3;1H
Formula Weight 215.0713
Percent Purity >95%
Chemical Name or Material (+/-)-Salsolinol, Hydrochloride
Grade HPLC