CAS RN 79923-51-6
CAS RN 79923-51-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
rac Salsolinol Hydrochloride, TRC
CAS: 79923-51-6 Molecular Formula: C10 H13 N O2 . Cl H Molecular Weight (g/mol): 215.68 IUPAC Name: 1-methyl-1,2,3,4-tetrahydroisoquinoline-6,7-diol;hydrochloride SMILES: Cl.CC1NCCc2cc(O)c(O)cc12