missing translation for 'onlineSavingsMsg'
Learn More

rac Albuterol, TRC

Product Code. 30137317
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137317 50 mg 50mg
30135319 100 mg 100mg
17903925 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137317 Supplier Toronto Research Chemicals Supplier No. TRCA51450150MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18559-94-9
Molecular Formula C13 H21 N O3
Molecular Weight (g/mol) 239.31
Synonym 1,3-Benzenedimethanol, α1-[[(1,1-dimethylethyl)amino]methyl]-4-hydroxy-, m-Xylene-α,α'-diol, α1-[(tert-butylamino)methyl]-4-hydroxy- (8CI), (±)-Albuterol, (±)-Salbutamol, 1-(tert-Butylaminomethyl)-4-hydroxy-m-xylene-α1,α3-diol, 2-(tert-Butylamino)-1-[(4-hydroxy-3-hydroxymethyl)phenyl]ethanol, 4-Hydroxy-3-(hydroxymethyl)-α-[[(1,1-dimethylethyl)amino]methyl]benzenemethanol, AH 3365, Airomir, Albuterol, DL-N-tert-Butyl-2(4-hydroxy-3-hydroxymethylphenyl)2-hydroxyethylamine, Eolene, Etinoline, Proventil HFA, Proventil Inhaler, Sabumol, Salamol EB, Salbutamol, Saventol, Spreor, Sultanol, Sultanol N, Ventalin Inhaler, Ventolin, Ventoline, Volmax, dl-Albuterol, dl-Salbutamol, m-Xylene α,α'-diol, α-[(tert-butylamino)methyl]-4-hydroxy-, α1-tert-Butylaminomethyl-4-hydroxy-m-xylene-α1,α3-diol
IUPAC Name 4-[2-(tert-butylamino)-1-hydroxyethyl]-2-(hydroxymethyl)phenol
SMILES CC(C)(C)NCC(O)c1ccc(O)c(CO)c1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C13H21NO3/c1-13(2,3)14-7-12(17)9-4-5-11(16)10(6-9)8-15/h4-6,12,14-17H,7-8H2,1-3H3
Formula Weight 239.1521
Percent Purity >95%
Chemical Name or Material Salbutamol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.