accessibility menu, dialog, popup

UserName

CAS RN 18559-94-9

H3CH3CCH3NHOHOHHO

CAS RN 18559-94-9

IUPAC Name: 4-[2-(tert-butylamino)-1-hydroxyethyl]-2-(hydroxymethyl)phenol
Synonyms: salbutamol ventolin hfa albuterol proventil xopenex hfa ProAir HFA
Molecular Weight (g/mol): 239.32
Molecular Formula: C13H21NO3
InChi Key: NDAUXUAQIAJITI-UHFFFAOYNA-N
SMILES: CC(C)(C)NCC(O)C1=CC=C(O)C(CO)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results