missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Mouse IgG3 Heavy Chain Secondary Antibody, Novus Biologicals™
Shop All Bio Techne ProductsDescription
IgG3 Heavy Chain Polyclonal specifically detects IgG3 Heavy Chain in Mouse samples. It is validated for ELISA, Flow Cytometry, Fluorophore-linked immunosorbent assay, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Western Blot.
Specifications
Specifications
| Antigen | IgG3 Heavy Chain |
| Applications | ELISA, Flow Cytometry, Fluorophore-Linked Immunosorbent Assay, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000 - 1:5000, Flow Cytometry, ELISA 1:10000 - 1:50000, Immunohistochemistry 1:1000 - 1:5000, Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay |
| Gene Alias | DKFZp686H11213, FLJ39988, FLJ40036, FLJ40253, FLJ40587, FLJ40789, FLJ40834, IgG3, immunoglobulin heavy constant gamma 3 (G3m marker), MGC45809 |
| Gene Symbols | IGHG3 |
| Host Species | Rabbit |
| Immunogen | Mouse IgG3 heavy chain |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?