missing translation for 'onlineSavingsMsg'
Learn More

Rabbit anti-Mouse IgG3 Heavy Chain Secondary Antibody, DyLight 488, Novus Biologicals™

Product Code. 18215005 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.5 mL
Unit Size:
0.5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18215005 0.5 mL 0.5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18215005 Supplier Novus Biologicals Supplier No. NBP172796G0.5ML
  • ONLINE EXCLUSIVE PRICE
    504.00 EUR
    476.70 EUR / 0.5mL
    Save 27.30 EUR
    5% Off
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IgG3 Polyclonal specifically detects IgG3 in Mouse samples. It is validated for ELISA, Flow Cytometry, Fluorophore-linked immunosorbent assay, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG3
Applications ELISA, Flow Cytometry, Fluorophore-Linked Immunosorbent Assay, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate DyLight 488
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50 mM Sodium Borate
Gene Alias DKFZp686H11213, FLJ39988, FLJ40036, FLJ40253, FLJ40587, FLJ40789, FLJ40834, IgG3, immunoglobulin heavy constant gamma 3 (G3m marker), MGC45809
Gene Symbols IGHG3
Host Species Rabbit
Immunogen Mouse IgG3 heavy chain
Purification Method Affinity Purified
Quantity 0.5 mL
Research Discipline Immunology
Gene ID (Entrez) 3502
Test Specificity Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Rabbit Serum, Mouse Serum and Mouse IgG.
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.