Learn More
(R,S)-Anatabine, TRC
Product Code. 18042310
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18042310 10 mg 10mg
18013320 100 mg 100mg
18022920 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 18042310 Supplier Toronto Research Chemicals Supplier No. TRCA63750010MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2743-90-0
Molecular Formula C10 H12 N2
Molecular Weight (g/mol) 160.22
Synonym 2,3'-Bipyridine, 1,2,3,6-tetrahydro- (9CI, ACI), 1,2,3,6-Tetrahydro-2,3'-bipyridine (ACI), 2,3'-Bipyridine, 1,2,3,6-tetrahydro-, (±)- (ZCI), (RS)-Anatabine, (±)-1,2,3,6-Tetrahydro-2,3'-bipyridine, (±)-Anatabine, 1,2,3,6-Tetrahydro-2,3'-dipyridine, 3-(1,2,3,6-Tetrahydropyridin-2-yl)pyridine
IUPAC Name 3-(1,2,3,6-tetrahydropyridin-2-yl)pyridine
SMILES C1NC(CC=C1)c2cccnc2
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C10H12N2/c1-2-7-12-10(5-1)9-4-3-6-11-8-9/h1-4,6,8,10,12H,5,7H2
Formula Weight 160.1
Chemical Name or Material (+/-)-Anatabine
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.