CAS RN 2743-90-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
(2RS)-1,2,3,6-Tetrahydro-2,3'-bipyridyl ((RS)-Anatabine), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
(R,S)-Anatabine, TRC
CAS: 2743-90-0 Molecular Formula: C10 H12 N2 Molecular Weight (g/mol): 160.22 Synonym: 2,3'-Bipyridine, 1,2,3,6-tetrahydro- (9CI, ACI),1,2,3,6-Tetrahydro-2,3'-bipyridine (ACI),2,3'-Bipyridine, 1,2,3,6-tetrahydro-, (±)- (ZCI),(RS)-Anatabine,(±)-1,2,3,6-Tetrahydro-2,3'-bipyridine,(±)-Anatabine,1,2,3,6-Tetrahydro-2,3'-dipyridine,3-(1,2,3,6-Tetrahydropyridin-2-yl)pyridine IUPAC Name: 3-(1,2,3,6-tetrahydropyridin-2-yl)pyridine SMILES: C1NC(CC=C1)c2cccnc2