Learn More
(R)-Anabasine, >98% ee, TRC
Product Code. 30134692
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
10mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134692 2.5 mg 2.5mg
30133260 5 mg 5mg
18040320 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30134692 Supplier Toronto Research Chemicals Supplier No. TRCA6371652.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 34366-21-7
Molecular Formula C10 H14 N2
Molecular Weight (g/mol) 162.23
Synonym Pyridine, 3-(2R)-2-piperidinyl-, 2αH-Anabasine (8CI), Pyridine, 3-(2-piperidinyl)-, (R)-, 3-(2R)-2-Piperidinylpyridine, (+)-Anabasine, (2'R)-Anabasine, (R)-Anabasine
IUPAC Name 3-[(2R)-piperidin-2-yl]pyridine
SMILES C1CC[C@@H](NC1)c2cccnc2
Specifications
Analyte or Component Names (R)-Anabasine
Shipping Condition Room Temperature
InChI Formula InChI=1S/C10H14N2/c1-2-7-12-10(5-1)9-4-3-6-11-8-9/h3-4,6,8,10,12H,1-2,5,7H2/t10-/m1/s1
Solution Type Neat
Industry Type Forensics
Quantity 2.5 mg
Formula Weight 162.1157
Chemical Name or Material (R)-Anabasine