CAS RN 34366-21-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(R)-Anabasine, >98% ee, TRC
CAS: 34366-21-7 Molecular Formula: C10 H14 N2 Molecular Weight (g/mol): 162.23 Synonym: Pyridine, 3-(2R)-2-piperidinyl-,2αH-Anabasine (8CI),Pyridine, 3-(2-piperidinyl)-, (R)-,3-(2R)-2-Piperidinylpyridine,(+)-Anabasine,(2'R)-Anabasine,(R)-Anabasine IUPAC Name: 3-[(2R)-piperidin-2-yl]pyridine SMILES: C1CC[C@@H](NC1)c2cccnc2