missing translation for 'onlineSavingsMsg'
Learn More

(R)-1-(Boc-amino)-2-propanol, 98%

Product Code. 15443657
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15443657 5 g 5g
15453657 250 mg 250mg
11372359 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15443657 Supplier Thermo Scientific Alfa Aesar Supplier No. H27296.06
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 119768-44-4 | C8H17NO3 | 175.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119768-44-4
Molecular Formula C8H17NO3
Molecular Weight (g/mol) 175.23
MDL Number MFCD04974338
InChI Key YNJCFDAODGKHAV-ZCFIWIBFSA-N
Synonym n-boc-r-1-amino-2-propanol, r-tert-butyl 2-hydroxypropyl carbamate, r-1-boc-amino-2-propanol, tert-butyl n-2r-2-hydroxypropyl carbamate, n-boc-1-amino-2-propanol, n-boc-r-isopropanolamine, t-butyl 2r-2-hydroxypropylcarbamate, r-tert-butyl 2-hydroxypropylcarbamate
PubChem CID 12094522
IUPAC Name tert-butyl N-[(2R)-2-hydroxypropyl]carbamate
SMILES C[C@@H](O)CNC(=O)OC(C)(C)C

Specifications

Refractive Index 1.446
Quantity 5 g
Formula Weight 175.23
Optical Rotation −24° (c=1 in Chloroform)
Percent Purity 98%
Chemical Name or Material (R)-1-(Boc-amino)-2-propanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.