accessibility menu, dialog, popup

UserName

CAS RN 119768-44-4

IUPAC Name: tert-butyl N-[(2R)-2-hydroxypropyl]carbamate
Synonyms: tert-butyl N-[(2R)-2-hydroxypropyl]carbamate
Molecular Weight (g/mol): 175.23
Molecular Formula: C8H17NO3
InChi Key: YNJCFDAODGKHAV-ZCFIWIBFSA-N
SMILES: C[C@@H](O)CNC(=O)OC(C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results