Learn More
(R)-1-(Boc-amino)-2-propanol, 98%
Product Code. 11372359
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11372359 1 g 1g
15453657 250 mg 250mg
15443657 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11372359 Supplier Thermo Scientific Alfa Aesar Supplier No. H27296.03
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
CAS: 119768-44-4 | C8H17NO3 | 175.23 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 119768-44-4
Molecular Formula C8H17NO3
Molecular Weight (g/mol) 175.23
MDL Number MFCD04974338
InChI Key YNJCFDAODGKHAV-ZCFIWIBFSA-N
Synonym n-boc-r-1-amino-2-propanol, r-tert-butyl 2-hydroxypropyl carbamate, r-1-boc-amino-2-propanol, tert-butyl n-2r-2-hydroxypropyl carbamate, n-boc-1-amino-2-propanol, n-boc-r-isopropanolamine, t-butyl 2r-2-hydroxypropylcarbamate, r-tert-butyl 2-hydroxypropylcarbamate
PubChem CID 12094522
IUPAC Name tert-butyl N-[(2R)-2-hydroxypropyl]carbamate
SMILES C[C@@H](O)CNC(=O)OC(C)(C)C
Specifications
Refractive Index 1.446
Quantity 1 g
Formula Weight 175.23
Optical Rotation −24° (c=1 in Chloroform)
Percent Purity 98%
Chemical Name or Material (R)-1-(Boc-amino)-2-propanol