missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-1,2-Bis(diphenylphosphino)propane, 98%, Thermo Scientific™

Product Code. 11494527
Change view
Click to view available options
Quantity:
0.25 g
1 g
Unit Size:
0.25g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11494527 0.25 g 0.25g
11484527 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11494527 Supplier Thermo Scientific Alfa Aesar Supplier No. B21078.01
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67884-32-6
Molecular Formula C27H26P2
Molecular Weight (g/mol) 412.45
MDL Number MFCD00012309
InChI Key WGOBPPNNYVSJTE-UHFFFAOYNA-N
Synonym r-+-1,2-bis diphenylphosphino propane, r-prophos, r-+-bis-1,2-diphenylphosphino propane, r-propane-1,2-diylbis diphenylphosphine, r-+-prophos, r-+-1,2bis diphenylphosphino propane, r-+-bis 1,2-diphenylphosphino propane, r-+-propylenebis 1,2-diphenylphosphine
PubChem CID 2724539
IUPAC Name [(2R)-1-diphenylphosphanylpropan-2-yl]-diphenylphosphane
SMILES CC(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 71°C to 73°C
Quantity 0.25 g
Sensitivity Air Sensitive
Formula Weight 412.46
Percent Purity 98%
Chemical Name or Material (R)-(+)-1,2-Bis(diphenylphosphino)propane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.