CAS RN 67884-32-6
IUPAC Name:
[1-(diphenylphosphanyl)propan-2-yl]diphenylphosphane
Synonyms:
[1-(diphenylphosphanyl)propan-2-yl]diphenylphosphane
Molecular Weight (g/mol):
412.45
Molecular Formula:
C27H26P2
InChi Key:
WGOBPPNNYVSJTE-UHFFFAOYNA-N
SMILES:
CC(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
(R)-(+)-1,2-Bis(diphenylphosphino)propane, 98%, Thermo Scientific™
CAS: 67884-32-6 Molecular Formula: C27H26P2 Molecular Weight (g/mol): 412.45 MDL Number: MFCD00012309 InChI Key: WGOBPPNNYVSJTE-UHFFFAOYNA-N Synonym: r-+-1,2-bis diphenylphosphino propane,r-prophos,r-+-bis-1,2-diphenylphosphino propane,r-propane-1,2-diylbis diphenylphosphine,r-+-prophos,r-+-1,2bis diphenylphosphino propane,r-+-bis 1,2-diphenylphosphino propane,r-+-propylenebis 1,2-diphenylphosphine PubChem CID: 2724539 IUPAC Name: [(2R)-1-diphenylphosphanylpropan-2-yl]-diphenylphosphane SMILES: CC(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1