missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ QS 11
GREENER_CHOICE

Product Code. 18055342
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18055342 10 mg 10mg
18035732 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18055342 Supplier Tocris Bioscience™ Supplier No. 3324/10
  • ONLINE EXCLUSIVE PRICE
    310.00 EUR
    292.95 EUR / 10mg
    Save 17.05 EUR
    5% Off
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

ARFGAP1 inhibitor; modulates Wnt/β-catenin signaling

GTPase activating protein of ADP-ribosylation factor 1 (ARFGAP1) inhibitor. Modulates ARF-GTP levels and synergizes with the Wnt/β-catenin signaling pathway to upregulate β-catenin nuclear translocation. Also reduces in vitro migration of metastatic human breast cancer cells.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 944328-88-5
Molecular Formula C36H33N5O2

Specifications

Quantity 10 mg
Formula Weight 567.68
Purity >98%
Molecular Weight (g/mol) 567.693
Chemical Name or Material QS 11
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.