missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-2,3-dicarboxylic anhydride, 98%

Product Code. 11416157
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11416157 5 g 5g
11426157 25 g 25g
11436157 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11416157 Supplier Thermo Scientific Alfa Aesar Supplier No. B25118.06
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 699-98-9 | C7H3NO3 | 149.105 g/mol

Pyridine-2,3-dicarboxylic anhydride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyridine-2,3-dicarboxylic anhydride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Optimal solubility in water.

Notes
Moisture sensitive. Store away from moisture and oxidizing agents under dry inert gas. Protect from humidity and water. Incompatible with oxidizing agents and moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 699-98-9
Molecular Formula C7H3NO3
Molecular Weight (g/mol) 149.105
MDL Number MFCD00005915
InChI Key MCQOWYALZVKMAR-UHFFFAOYSA-N
Synonym 2,3-pyridinedicarboxylic anhydride, furo 3,4-b pyridine-5,7-dione, quinolinic anhydride, 2,3-pyridinedicarboxylicanhydride, pyridine-2,3-dicarboxylic anhydride, 5h,7h-furo 3,4-b pyridine-5,7-dione, furano 3,4-b pyridine-5,7-dione, pyridine-2,3-dicarboxylic acid anhydride, 2,3-pyridinedicarboxylic anhydride quinolinic anhydride
PubChem CID 69688
IUPAC Name furo[3,4-b]pyridine-5,7-dione
SMILES C1=CC2=C(C(=O)OC2=O)N=C1

Specifications

Melting Point 134°C to 138°C
Quantity 5 g
Beilstein 125111
Sensitivity Moisture sensitive
Solubility Information Optimal solubility in water.
Formula Weight 149.11
Percent Purity 98%
Chemical Name or Material Pyridine-2,3-dicarboxylic anhydride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.