CAS RN 699-98-9
CAS RN 699-98-9
IUPAC Name:
5H,7H-furo[3,4-b]pyridine-5,7-dione
Synonyms:
furo[3,4-b]pyridine-5,7-dione
Molecular Weight (g/mol):
149.11
Molecular Formula:
C7H3NO3
InChi Key:
MCQOWYALZVKMAR-UHFFFAOYSA-N
SMILES:
O=C1OC(=O)C2=NC=CC=C12
1
–
1
of
1
results
1
Pyridine-2,3-dicarboxylic anhydride, 98%
CAS: 699-98-9 Molecular Formula: C7H3NO3 Molecular Weight (g/mol): 149.105 MDL Number: MFCD00005915 InChI Key: MCQOWYALZVKMAR-UHFFFAOYSA-N Synonym: 2,3-pyridinedicarboxylic anhydride,furo 3,4-b pyridine-5,7-dione,quinolinic anhydride,2,3-pyridinedicarboxylicanhydride,pyridine-2,3-dicarboxylic anhydride,5h,7h-furo 3,4-b pyridine-5,7-dione,furano 3,4-b pyridine-5,7-dione,pyridine-2,3-dicarboxylic acid anhydride,2,3-pyridinedicarboxylic anhydride quinolinic anhydride PubChem CID: 69688 IUPAC Name: furo[3,4-b]pyridine-5,7-dione SMILES: C1=CC2=C(C(=O)OC2=O)N=C1