missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PTPRN2 293T Cell Transient Overexpression Lysate
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.01mL
Specifications
Specifications
| Quantity | 0.1 mL |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| Research Category | Lipid and Metabolism, Protein Phosphatase |
| Target | PTPRN2 |
| Synonym | EC 3.1.3.48, IA-2beta, IAR/receptor-like protein-tyrosine phosphatase, ICAARIARPTPRP, Islet cell autoantigen-related protein, KIAA0387, phogrin, protein tyrosine phosphatase receptor pi, protein tyrosine phosphatase, receptor type, N polypeptide 2, receptor-type tyrosine-protein phosphatase N2, R-PTP-N2, tyrosine phosphatase IA-2 beta |
| Gene Symbol | PTPRN2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction