missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PTPN23 293T Cell Transient Overexpression Lysate
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.01mL
Specifications
Specifications
| Quantity | 0.1 mL |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| Target | PTPN23 |
| Synonym | EC 3.1.3.48, HDPTP, HD-PTPDKFZp564F0923, His domain-containing protein tyrosine phosphatase, His-domain protein tyrosine phosphatase, KIAA1471DKFZP564F0923, Protein tyrosine phosphatase TD14, protein tyrosine phosphatase, non-receptor type 23, PTP-TD14, tyrosine-protein phosphatase non-receptor type 23 |
| Gene Symbol | PTPN23 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction