missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ PSAT1 Protein

Product Code. 18485850 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.5 mg
100μg
Unit Size:
0.50mg
100µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18485850 0.5 mg 0.50mg
18214979 100μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18485850 Supplier Novus Biologicals™ Supplier No. NBP1788270.5mg
  • ONLINE EXCLUSIVE PRICE
    1600.00 EUR
    1509.90 EUR / 0.5mg
    Save 90.10 EUR
    5% Off
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-370 of Human PSAT1 The Recombinant Human PSAT1 Protein is derived from E. coli. The Recombinant Human PSAT1 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_478059
Concentration 1 mg/mL
For Use With (Application) SDS-PAGE
Formulation Liquid. 20 mM Tris-HCl buffer (pH 8.0) containing 20% glycerol, 1 mM DTT
Gene ID (Entrez) 29968
Molecular Weight (g/mol) 43 kDa
Purification Method SDS-PAGE
Quantity 0.5 mg
Source E. coli
Immunogen PSAT1, 1-370 aa, Human, His tag, E. coli (NP_478059).
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name PSAT1
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.