missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Protein Tyrosine Phosphatase Activity Assay Kit (Fluorometric)
Shop All Bio Techne Products
Click to view available options
Quantity:
100 Assays
Unit Size:
100 assays
Description
Detection method: Fluorescence (Ex/Em = 368/440 nm). Applications: Rapid assessment of PTPase activity in biological samples or recombinant PTPase preparations.
Kit Contents:
- PTPase Assay Buffer
- PTPase Inhibitor (Suramin)
- PTPase Positive Control
- PTPase Substrate
- Fluorescent Standard
- Disulfide Reducing Agent (DTT)
Specifications
Specifications
| Quantity | 100 Assays |
| Product Type | Activity Assay Kit (Fluorometric) |
| Content And Storage | Store at -20°C in the dark. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction