Learn More
Cytiva Protein G Sepharose™ 4 Fast Flow Media
Product Code. 11524935 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
200 mL
Unit Size:
200mL
25mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11524935 5 mL 5mL
11534935 25 mL 25mL
11544935 200 mL 200mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11524935 Supplier Cytiva Supplier No. 17061801
 Restricted Item
Estimated Shipment: 10-12-2026
Rapid processing of large volumes, recoveries of 70 to 90%. Cytiva Protein G Sepharose™ 4 Fast Flow Media is recombinant protein G coupled to Sepharose 4 Fast Flow.
  • Binding specificities that complement Protein A media
  • Binds a broad range of IgG species and subclasses
  • High capacity, >20mg human IgG/mL medium; high flowrate
  • Multi-point attachment minimizes ligand leakage
  • Used in a range of diagnostic and research applications
TRUSTED_SUSTAINABILITY
Specifications
Ligand Concentration 2mg Protein G/mL Medium
Matrix 4% Cross Linked Agarose
Particle Size 90 μm
pH 3 to 9
Quantity 5 mL
Content And Storage 2°C to 8°C
For Use With (Application) Used in a range of diagnostic and research applications
Ligand Recombinant Protein G lacking albumin binding region
compliance-icons
Product Identifier
  • Protein G Sepharose 4 Fast Flow, 5 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.