Learn More
Probucol, Thermo Scientific™
Product Code. 17730659 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17730659 25 g 25g
17720659 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17730659 Supplier Thermo Scientific Acros Supplier No. 469490250
 Restricted Item
In Stock
Add to basket
Specifications
Chemical Name or Material Probucol
CAS 23288-49-5
Synonym 4,4 '-(Isopropylidendithio)bis[2,6-di-tert-butylphenol]
Quantity 25 g
Solubility insoluble aqueous solution: soluble in methanol, acetone, acetonitrile, THF
Melting Point 125°C to 128°C
Shipping Condition No special requirements
Product Line Life science
Molecular Formula C31 H48 O2 S2
Physical Form Powder or Crystalline Powder
Color White to Almost-white
Purity ≥97.5%