Learn More
Potassium Trichloroammineplatinate, TRC
Product Code. 30425146
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30425146 1 g
30421284 100 mg
30423387 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30425146 Supplier Toronto Research Chemicals Supplier No. TRCP6985501G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 13820-91-2
Molecular Formula K . Cl3 H3 N Pt
Molecular Weight (g/mol) 357.57
Synonym Platinate(1-), amminetrichloro-, potassium, (SP-4-2)- (9CI), Platinate(1-), amminetrichloro-, potassium (8CI), Potassium trichloroammineplatinate(II) (6CI,7CI), NSC 123898, Potassium amminetrichloroplatinate, Potassium amminetrichloroplatinate(1-) Coordination Compound
SMILES [K+].[Cl-][Pt+2]([Cl-])([Cl-])[NH3]
Specifications
Quantity 1 g
InChI Formula InChI=1S/3ClH.K.H3N.Pt/h3*1H;;1H3;/q;;;+1;;+2/p-3
Formula Weight 355.8616
Chemical Name or Material Potassium Trichloroammineplatinate