CAS RN 13820-91-2
CAS RN 13820-91-2
IUPAC Name:
potassium trichloroplatinumuide amine
Synonyms:
potassium trichloroplatinumuide amine
Molecular Weight (g/mol):
357.56
Molecular Formula:
Cl3H3KNPt
InChi Key:
PIFYEDQIQOTXLO-UHFFFAOYSA-K
SMILES:
N.[K+].Cl[Pt-](Cl)Cl
1
–
2
of
2
results
2
Potassium Trichloroammineplatinate, TRC
CAS: 13820-91-2 Molecular Formula: K . Cl3 H3 N Pt Molecular Weight (g/mol): 357.57 Synonym: Platinate(1-), amminetrichloro-, potassium, (SP-4-2)- (9CI),Platinate(1-), amminetrichloro-, potassium (8CI),Potassium trichloroammineplatinate(II) (6CI,7CI),NSC 123898,Potassium amminetrichloroplatinate,Potassium amminetrichloroplatinate(1-) Coordination Compound SMILES: [K+].[Cl-][Pt+2]([Cl-])([Cl-])[NH3]