missing translation for 'onlineSavingsMsg'
Learn More

Potassium pentachloronitrosylruthenate(II), Ru 25.8%, Thermo Scientific Chemicals

Product Code. 11346048
Change view
Click to view available options
Quantity:
0.25 g
1 g
Unit Size:
0.25g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11346048 0.25 g 0.25g
11336048 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11346048 Supplier Thermo Scientific Alfa Aesar Supplier No. 010534.01
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14854-54-7 | Molecular Formula: Cl5K2NORu | Molecular Weight: 386.52 g/mol

Used as precipitation agent in Heterobimetallic coordination polymers involving 3d metal complexes and heavier transition metals cyanometallates. Ruthenium complexes are presently an object of great attention in the field of medicinal chemistry, as antitumor agents with selective antimetastatic properties and low systemic toxicity. Ruthenium compounds appear to penetrate reasonably well the tumor cells and bind effectively to DNA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14854-54-7
Molecular Formula Cl5K2NORu
Molecular Weight (g/mol) 386.52
MDL Number MFCD00049789
InChI Key ZLWZLQOGXCFPCK-UHFFFAOYSA-I
Synonym dipotassium nitrosylpentachlororuthenate ii, dipotassium pentachloro nitrosyl ruthenate ii, dipotassium pentachloro nitroso rutheniumdiuide, potassium pentachloronitrosylruthenate ii , ru
PubChem CID 71311320
SMILES [K+].[K+].Cl[Ru++](Cl)(Cl)(Cl)(Cl)N=O

Specifications

Melting Point 220°C (decomposition)
Physical Form Crystalline
Quantity 0.25 g
Solubility Information Soluble in water.
IUPAC Name dipotassium;nitroxyl anion;pentachlororuthenium(2-)
Formula Weight 386.55
Concentration or Composition (by Analyte or Components) Ru 25.8%
Chemical Name or Material Potassium pentachloronitrosylruthenate(II)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.