accessibility menu, dialog, popup

UserName

CAS RN 14854-54-7

KRuClClNClClClOK

CAS RN 14854-54-7

IUPAC Name: λ²-ruthenium(2+) dipotassium iminooxidanium pentachloride
Synonyms: λ²-ruthenium(2+) dipotassium nitrosonium pentachloride
Molecular Weight (g/mol): 386.52
Molecular Formula: Cl5K2NORu
InChi Key: FCWXUHRWZLOTGH-UHFFFAOYSA-I
SMILES: [Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[K+].[K+].[Ru++].N#[O+]
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results